Learn More
Medchemexpress LLC IL-17A modulator-1 | 2748749-29-1 | 98.80% | C33H31N5O4 | 50 MG
SDP
Catalog No. 5000354426
Encompass_Preferred
IL-17A modulator-1 is an IL-17A modulator extracted from patent WO2021239743+A1, specifically example 9. It functions by inhibiting the biological action of IL-17A, demonstrating a pIC50 of 8.2. This compound is intended for research purposes concerning diseases or disorders linked to the modulation of IL-17A activity.
  • Inhibits the biological action of IL-17A
  • Demonstrates a pIC50 of 8.2
  • Suitable for research in immune component conditions and autoimmune pathologies
  • Applicable for research in cancer and neurodegenerative disorders
  • Appears as a white to off-white solid
Catalog No. 50-003-54426 Supplier Medchemexpress LLC Supplier No. HY14543050MG
IL-17A modulator-1 is an IL-17A modulator extracted from patent WO2021239743+A1, specifically example 9. It functions by inhibiting the biological action of IL-17A, demonstrating a pIC50 of 8.2. This compound is intended for research purposes concerning diseases or disorders linked to the modulation of IL-17A activity.
  • Inhibits the biological action of IL-17A
  • Demonstrates a pIC50 of 8.2
  • Suitable for research in immune component conditions and autoimmune pathologies
  • Applicable for research in cancer and neurodegenerative disorders
  • Appears as a white to off-white solid