Learn More
Medchemexpress LLC IL-17A modulator-2 | 2748749-47-3 | 98.4% | 577.63 | C33H31N5O5 | 5 MG
SDP
Catalog No. 5000221482
Encompass_Preferred
IL-17A modulator-2 is a small-molecule research compound that inhibits the biological activity of interleukin-17A. Reported from patent WO2021239743 (example 27), it is intended for use in preclinical studies investigating immune-related, autoimmune, cancer, and neurodegenerative disorders.
  • Inhibits interleukin-17A with a reported pIC50 of 8.3.
  • High purity suitable for research applications (≈98.4%).
  • Molecular weight 577.63; formula C33H31N5O5.
  • Available in small quantities appropriate for in vitro assays.
  • Reportedly derived from a patent example, supporting reproducible synthesis.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-21482 Supplier Medchemexpress LLC Supplier No. HY1454295MG
IL-17A modulator-2 is a small-molecule research compound that inhibits the biological activity of interleukin-17A. Reported from patent WO2021239743 (example 27), it is intended for use in preclinical studies investigating immune-related, autoimmune, cancer, and neurodegenerative disorders.
  • Inhibits interleukin-17A with a reported pIC50 of 8.3.
  • High purity suitable for research applications (≈98.4%).
  • Molecular weight 577.63; formula C33H31N5O5.
  • Available in small quantities appropriate for in vitro assays.
  • Reportedly derived from a patent example, supporting reproducible synthesis.
  • Intended for research use only; not for human therapeutic use.