Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-17A modulator-2 | 2748749-47-3 | 98.4% | C33H31N5O5 | 25 MG
SDP

Catalog No. 5000353112
Encompass_Preferred

IL-17A modulator-2 is a chemical compound with the molecular formula C33H31N5O5, presented as a white to off-white solid. It has a molecular weight of 577.63 and demonstrates a purity of 98.44% (HPLC). This compound is ideal for research applications requiring a high-purity reagent, with specific storage recommendations to maintain its stability.

  • White to off-white solid appearance
  • Molecular formula: C33H31N5O5
  • Molecular weight: 577.63
  • Purity: 98.44% (HPLC)
  • Solid form storage: 4°C, sealed, away from moisture and light
  • In solvent storage: -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Catalog No. 50-003-53112 Supplier Medchemexpress LLC Supplier No. 25MGHY145429
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IL-17A modulator-2 is a chemical compound with the molecular formula C33H31N5O5, presented as a white to off-white solid. It has a molecular weight of 577.63 and demonstrates a purity of 98.44% (HPLC). This compound is ideal for research applications requiring a high-purity reagent, with specific storage recommendations to maintain its stability.

  • White to off-white solid appearance
  • Molecular formula: C33H31N5O5
  • Molecular weight: 577.63
  • Purity: 98.44% (HPLC)
  • Solid form storage: 4°C, sealed, away from moisture and light
  • In solvent storage: -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.