Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-17A modulator-2 | 2748749-47-3 | >98% | C33H31N5O5 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY14542950MG

Encompass_Preferred

IL-17A modulator-2 is an IL-17A modulator, extracted from patent WO2021239743+A1, example 27. It inhibits the biological action of IL-17A with a pIC50 of 8.3.

  • Chemical formula: C33H31N5O5
  • Molecular weight: 577.63
  • Shelf life: 1095 days
  • Solubility in DMSO: 100 mg/mL (173.12 mM); requires ultrasonic
  • Storage: 2-8°C, protected from light, desiccated

Catalog No. 50-003-55263


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.