Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Il-17a modulator-3 | 2467732-95-0 | 99.4% | 639.80 | C34H50FN7O4 | 10 MG
SDP

Catalog No. 5000287407
Encompass_Preferred

Research-grade small molecule for in vitro investigation of IL-17A-related biology, including inflammation, cancer, and autoimmune disease pathways. Supplied as a solid with documented physicochemical properties, solubility, and storage recommendations to support reproducible experimental use.

  • Modulates IL-17A activity in vitro, with reported IC50 < 10 μM.
  • High chemical purity suitable for research applications.
  • Molecular formula C34H50FN7O4 and molecular weight 639.80 Da.
  • Soluble in DMSO at high concentration; ultrasonic assistance recommended.
  • Recommended storage conditions defined for powder and solutions to preserve stability.

Catalog No. 50-002-87407 Supplier Medchemexpress LLC Supplier No. HY14827210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Research-grade small molecule for in vitro investigation of IL-17A-related biology, including inflammation, cancer, and autoimmune disease pathways. Supplied as a solid with documented physicochemical properties, solubility, and storage recommendations to support reproducible experimental use.

  • Modulates IL-17A activity in vitro, with reported IC50 < 10 μM.
  • High chemical purity suitable for research applications.
  • Molecular formula C34H50FN7O4 and molecular weight 639.80 Da.
  • Soluble in DMSO at high concentration; ultrasonic assistance recommended.
  • Recommended storage conditions defined for powder and solutions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.