Learn More
Medchemexpress LLC IL-17A Mouse CHO 2ug
SDP
Catalog No. 5000179598
Encompass_Preferred
IL-17A is a homodimeric cytokine and plays a critical role in host defense mechanisms against many bacterial and fungal pathogens as well as allergic and autoimmune responses IL-17A induces the production of antimicrobial peptides cytokines chemokines and matrix metalloproteinases[1][2] IL-17A Protein Mouse (CHO) is a recombinant mouse IL-17A protein and is expressed in CHO cells It consists of 133 amino acids (A26-A158)
Catalog No. 50-001-79598 Supplier Medchemexpress LLC Supplier No. HYP73722UG
IL-17A is a homodimeric cytokine and plays a critical role in host defense mechanisms against many bacterial and fungal pathogens as well as allergic and autoimmune responses IL-17A induces the production of antimicrobial peptides cytokines chemokines and matrix metalloproteinases[1][2] IL-17A Protein Mouse (CHO) is a recombinant mouse IL-17A protein and is expressed in CHO cells It consists of 133 amino acids (A26-A158)