Learn More
Medchemexpress LLC IL-17A Mouse E. co 100ug
SDP
Catalog No. 5000182448
Encompass_Preferred
IL-17A protein is an important effector cytokine that activates the NF-kappa-B and MAPkinase pathways through the IL17RA-IL17RC receptor complex to protect host tissues from microbial threats As a key Th17 cytokine IL-17A mediates neutrophil activation chemotaxis and contributes to germinal center formation IL-17A Protein Mouse (E coli) is the recombinant mouse-derived IL-17A protein expressed by E coli with tag free
Catalog No. 50-001-82448 Supplier Medchemexpress LLC Supplier No. HYP73174A100UG
IL-17A protein is an important effector cytokine that activates the NF-kappa-B and MAPkinase pathways through the IL17RA-IL17RC receptor complex to protect host tissues from microbial threats As a key Th17 cytokine IL-17A mediates neutrophil activation chemotaxis and contributes to germinal center formation IL-17A Protein Mouse (E coli) is the recombinant mouse-derived IL-17A protein expressed by E coli with tag free