Learn More
Enzo Life Sciences IL-17A (mouse), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2440025
Encompass_Preferred
Alternative name: Interleukin-17A, IL-17, CTLA-8, Cytotoxic T-lymphocyte-associated antigen 8. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 134/268 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4052