Learn More
Medchemexpress LLC IL-17A Rabbit sf9 50ug
SDP
Catalog No. 5000182449
Encompass_Preferred
IL-17A is a homodimeric cytokine and plays a critical role in host defense mechanisms against many bacterial and fungal pathogens as well as allergic and autoimmune responses IL-17A induces the production of antimicrobial peptides cytokines chemokines and matrix metalloproteinases[1][2] IL-17A Protein Rabbit (sf9 His) is a recombinant rabbit IL-17A protein with His tag at the N-terminus and is expressed in sf9 insect cells It consists of 130 amino acids (G24-A153)
Catalog No. 50-001-82449 Supplier Medchemexpress LLC Supplier No. HYP7317650UG
IL-17A is a homodimeric cytokine and plays a critical role in host defense mechanisms against many bacterial and fungal pathogens as well as allergic and autoimmune responses IL-17A induces the production of antimicrobial peptides cytokines chemokines and matrix metalloproteinases[1][2] IL-17A Protein Rabbit (sf9 His) is a recombinant rabbit IL-17A protein with His tag at the N-terminus and is expressed in sf9 insect cells It consists of 130 amino acids (G24-A153)