Learn More
Medchemexpress LLC IL-17A Rat CHO 250ug | 250ug
SDP
Catalog No. 5000194620
Encompass_Preferred
The IL-17A protein has important heterodimerization and homodimerization activities and is involved in a variety of processes including responses to glucocorticoid stimulation and the regulation of cell death and transcription IL-17A is present in the cytoplasm and extracellular space and has been implicated in diseases such as pancreatitis inflammatory bowel disease renal disease and periodontal disease and may serve as a biomarker IL-17A Protein Rat (CHO) is the recombinant rat-derived IL-17A protein expressed by CHO with tag free
Catalog No. 50-001-94620 Supplier Medchemexpress LLC Supplier No. HYP79326250UG
The IL-17A protein has important heterodimerization and homodimerization activities and is involved in a variety of processes including responses to glucocorticoid stimulation and the regulation of cell death and transcription IL-17A is present in the cytoplasm and extracellular space and has been implicated in diseases such as pancreatitis inflammatory bowel disease renal disease and periodontal disease and may serve as a biomarker IL-17A Protein Rat (CHO) is the recombinant rat-derived IL-17A protein expressed by CHO with tag free