Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-17B ELISA Kit

Catalog No. EHIL17BX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHIL17BX5 5 x 96 Tests
EHIL17B 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHIL17BX5 Supplier Invitrogen™ Supplier No. EHIL17BX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Interleukin 17B (Hu IL-17B) ELISA quantitates Hu IL-17B in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-17B. Principle of the method The Human IL-17B solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a T cell-derived cytokine that shares sequence similarity with IL17. This cytokine was reported to stimulate the release of TNF alpha (TNF) and IL1 beta (IL1B) from a monocytic cell line. Immunohistochemical analysis of several nerve tissues indicated that this cytokine is primarily localized to neuronal cell bodies.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHF5
Assay Range 0.3-100 ng/mL
Assay Sensitivity 0.3 ng/mL
Calibration Range 0.3 ng/mL to 100 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cytokines & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF-7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF,SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1110006O16Rik; 1700006N07Rik; Cytokine CX1; Cytokine Zcyto7; Cytokine-like protein ZCYTO7; gene for cytokine-like protein; IL17B; IL-17B; IL20; IL-20; Interleukin; interleukin 17B; interleukin 20; interleukin-17 beta; interleukin-17B; Interleukin20; Interleukin-20; neuronal interleukin-17 related factor; neuronal interleukin-17-related factor; NIRF; UNQ516/PRO1031; ZCYTO71110006O16Rik; 1700006N07Rik; Cytokine CX1; Cytokine Zcyto7; Cytokine-like protein ZCYTO7; gene for cytokine-like protein; IL17B; IL-17B; IL20; IL-20; Interleukin; interleukin 17B; interleukin 20; interleukin-17 beta; interleukin-17B; Interleukin20; Interleukin-20; neuronal interleukin-17 related factor; neuronal interleukin-17-related factor; NIRF; UNQ516/PRO1031; ZCYTO7; IL-17B; IL-20; NIRF; ZCYTO7
Gene ID (Entrez) 27190
Gene Symbol IL17B
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases IL-17B, Cytokine Zcyto7, Interleukin-20, IL-20, Neuronal interleukin-17-related factor
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym UNQ516/PRO1031, IL-17 B, IL-20, NIRF, ZCYTO7
Target IL17B
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.