Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-17C Protein, His,Avitag™
Catalog No. 501956972
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-17C, His,Avitag (ILC-H82E5) is expressed from human 293 cells (HEK293). It contains AA His 19 - Val 197 (Accession # Q9P0M4-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.4 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-17C, His,Avitag (Cat. No. ILC-H82E5) with a linear range of 0.5-13 ng/mL (QC tested).
Catalog No. 50-195-6972 Supplier ACROBiosystems Supplier No. ILCH82E525UG
Recombinant Protein;25UG;Biotinylated Human IL-17C, His,Avitag (ILC-H82E5) is expressed from human 293 cells (HEK293). It contains AA His 19 - Val 197 (Accession # Q9P0M4-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.4 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-17C, His,Avitag (Cat. No. ILC-H82E5) with a linear range of 0.5-13 ng/mL (QC tested).