Learn More
ACROBiosystems ACROBiosystems Human IL-17C Protein, His Tag
Supplier:  ACROBiosystems ILCH52H71MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-17C, His Tag (ILC-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 19 - Val 197 (Accession # Q9P0M4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.6 kDa. The protein migrates as 24-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17C, His Tag (Cat. No. ILC-H52H7) at 2 μg/mL (100 μL/well) can bind Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-195-6819