Learn More
ACROBiosystems ACROBiosystems Human IL-17F / IL-24 Protein, His Tag
Supplier:  ACROBiosystems ILFH42401MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-17F, His Tag (ILF-H4240) is expressed from human 293 cells (HEK293). It contains AA Arg 31 - Gln 163 (Accession # AAH70124.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7263