Learn More
Genscript Corporation IL-17R alpha/CD217 hFc Chimera
Supplier:  Genscript Corporation Z04500100
Encompass
Interleukin 17 (also known as CTLA 8) is a T cell expressed pleotropic cytokine IL 17 binds to IL 17 receptor (IL 17 R) which shares no homology with any known family of receptors While the expression of IL 17 is restricted to activated T cells the IL 17 R mRNA exhibits a broad tissue distribution and has been detected in virtually all cells and tissues tested The human IL 17 R gene was localized to chromosome 22
Catalog No. 50-000-59292