Learn More
Medchemexpress LLC IL-17RA protein, rat (HEK293, His) | 2 UG
SDP
Catalog No. 5000292809
Encompass_Preferred
Recombinant rat interleukin-17 receptor A (IL-17RA) extracellular domain produced in HEK293 cells and supplied with a C-terminal His tag. The construct corresponds to 320 amino acids (M1-P320) and is intended for research applications including binding assays, pathway studies, and antibody validation.
  • Produced in HEK293 cells with C-terminal His tag.
  • Comprises 320 amino acids corresponding to M1-P320.
  • Suitable for binding assays, signaling studies, and antibody validation.
  • Supplied in small-scale quantities for exploratory research.
  • Purity and storage details available on the certificate of analysis.
Catalog No. 50-002-92809 Supplier Medchemexpress LLC Supplier No. HYP748252UG
Recombinant rat interleukin-17 receptor A (IL-17RA) extracellular domain produced in HEK293 cells and supplied with a C-terminal His tag. The construct corresponds to 320 amino acids (M1-P320) and is intended for research applications including binding assays, pathway studies, and antibody validation.
  • Produced in HEK293 cells with C-terminal His tag.
  • Comprises 320 amino acids corresponding to M1-P320.
  • Suitable for binding assays, signaling studies, and antibody validation.
  • Supplied in small-scale quantities for exploratory research.
  • Purity and storage details available on the certificate of analysis.