Learn More
Medchemexpress LLC IL-17RD Mouse CHO 2ug
SDP
Catalog No. 5000181566
Encompass_Preferred
Interleukin-17 receptor D (IL-17RD) also known as Sef (similar expression to fibroblast growth factor) is a type I transmembrane protein that can regulate several signaling pathways Within the cell IL-17RD expression has been localized mainly to the plasma membrane IL-17RD is a feedback inhibitor of fibroblast growth factor (FGF) signaling[1] IL-17RD Protein Mouse (CHO) is produced in CHO cells and consists of 2713 amino acids (G29-R299)
Catalog No. 50-001-81566 Supplier Medchemexpress LLC Supplier No. HYP727942UG
Interleukin-17 receptor D (IL-17RD) also known as Sef (similar expression to fibroblast growth factor) is a type I transmembrane protein that can regulate several signaling pathways Within the cell IL-17RD expression has been localized mainly to the plasma membrane IL-17RD is a feedback inhibitor of fibroblast growth factor (FGF) signaling[1] IL-17RD Protein Mouse (CHO) is produced in CHO cells and consists of 2713 amino acids (G29-R299)