Learn More
ACROBiosystems ACROBiosystems Human IL-17 RE (155-454) Protein, Fc Tag
Catalog No. 501956644
Encompass_Preferred
Recombinant Protein;1MG;Human IL-17 RE (155-454), Fc Tag (ILE-H5256) is expressed from human 293 cells (HEK293). It contains AA Thr 155 - His 454 (Accession # Q8NFR9-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17C, His Tag (Cat. No. ILC-H52H7) at 2 μg/mL (100 μL/well) can bind Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-195-6644 Supplier ACROBiosystems Supplier No. ILEH52561MG
Recombinant Protein;1MG;Human IL-17 RE (155-454), Fc Tag (ILE-H5256) is expressed from human 293 cells (HEK293). It contains AA Thr 155 - His 454 (Accession # Q8NFR9-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17C, His Tag (Cat. No. ILC-H52H7) at 2 μg/mL (100 μL/well) can bind Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) with a linear range of 1-10 ng/mL (QC tested).