Learn More
Medchemexpress LLC IL-18 Human N-His 100ug | 100ug
SDP
Catalog No. 5000184521
Encompass_Preferred
IL-18 protein is a pro-inflammatory cytokine that plays a critical role in epithelial barrier repair and polarized immune responses involving Th1 cells and NK cells After binding to IL18R1 and IL18RAP it forms a signaling ternary complex that activates NF-kappa-B leading to the synthesis of inflammatory mediators IL-18 Protein Human (N-His) is the recombinant human-derived IL-18 protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-84521 Supplier Medchemexpress LLC Supplier No. HYP70591A100UG
IL-18 protein is a pro-inflammatory cytokine that plays a critical role in epithelial barrier repair and polarized immune responses involving Th1 cells and NK cells After binding to IL18R1 and IL18RAP it forms a signaling ternary complex that activates NF-kappa-B leading to the synthesis of inflammatory mediators IL-18 Protein Human (N-His) is the recombinant human-derived IL-18 protein expressed by E coli with N-6 His labeled tag