Learn More
Medchemexpress LLC IL-18 Mouse soluti 100ug
SDP
Catalog No. 5000181317
Encompass_Preferred
IL-18 protein is a pro-inflammatory cytokine that plays an important role in epithelial barrier repair and immune response regulation by polarizing T helper 1 (Th1) cells and natural killer (NK) cells After binding to IL18R1 and IL18RAP receptors IL-18 forms a signaling ternary complex activates NF- -B and induces inflammatory mediators IL-18 Protein Mouse (solution) is the recombinant mouse-derived IL-18 protein expressed by E coli with tag free
Catalog No. 50-001-81317 Supplier Medchemexpress LLC Supplier No. HYP73181100UG
IL-18 protein is a pro-inflammatory cytokine that plays an important role in epithelial barrier repair and immune response regulation by polarizing T helper 1 (Th1) cells and natural killer (NK) cells After binding to IL18R1 and IL18RAP receptors IL-18 forms a signaling ternary complex activates NF- -B and induces inflammatory mediators IL-18 Protein Mouse (solution) is the recombinant mouse-derived IL-18 protein expressed by E coli with tag free