Learn More
Medchemexpress LLC IL-18BP Human CHO 10ug
SDP
Catalog No. 5000195477
Encompass_Preferred
IL-18BP Protein, Human (CHO) is a recombinant human interleukin-18 binding protein produced in CHO cells and supplied as a 10 μg research reagent. It neutralizes IL-18 activity and modulates Th1 cytokine responses; the product is characterized by defined biological activity and high purity for use in biochemical and cell-based studies.
  • Recombinant human IL-18 binding protein expressed in CHO cells.
  • High purity (>95%) as assessed by reducing SDS-PAGE.
  • Defined molecular weight (~42-44 kDa) under reducing conditions.
  • Demonstrated biological activity (ED50 < 30 ng/mL in KG-1 assay).
  • Supplied in a 10 μg format for small-scale experiments.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term use.
Catalog No. 50-001-95477 Supplier Medchemexpress LLC Supplier No. HYP721110UG
IL-18BP Protein, Human (CHO) is a recombinant human interleukin-18 binding protein produced in CHO cells and supplied as a 10 μg research reagent. It neutralizes IL-18 activity and modulates Th1 cytokine responses; the product is characterized by defined biological activity and high purity for use in biochemical and cell-based studies.
  • Recombinant human IL-18 binding protein expressed in CHO cells.
  • High purity (>95%) as assessed by reducing SDS-PAGE.
  • Defined molecular weight (~42-44 kDa) under reducing conditions.
  • Demonstrated biological activity (ED50 < 30 ng/mL in KG-1 assay).
  • Supplied in a 10 μg format for small-scale experiments.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term use.