Learn More
Enzo Life Sciences IL-19 (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2450100
Encompass_Preferred
Alternative name: Interleukin-19, Melanoma differentiation associated-like protein. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 154 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4055