Learn More
AdipoGen anti-IL-1alpha (mouse), mAb (Bamboo-2) (100 µg)
SDP
Supplier:  AdipoGen AG20B0058C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse IL-1alpha. Isotype: Mouse IgG1kappa. Clone: Bamboo-2. Applications: ELISA. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. IL-1alpha lacks a signal peptide and is secreted by an unconventional, endoplasmic reticulum-Golgi-independent mechanism. IL-1alpha was reported to be more widely and constitutively expressed and has intracellular functions, but also acts locally in a membrane-bound form by activating IL-1R1. Additionally, passive release of IL-1alpha upon cell death can trigger a sterile inflammatory response to dying cells. The cleavage of IL-1alpha is not mediated by caspase-1 and is not required for binding to IL-1R1. Recently it has been observed that all activators of the inflammasome NLRP3 induce the simultaneous secretion of IL-1alpha and IL-1beta. Although most activators fully rely on the inflammasome for IL-1alpha secretion, some induce the processing and secretion of IL-1alpha in an inflammasome-independent manner.
Catalog No. 50-205-2971