Learn More
Enzo Life Sciences IL-1alpha (human), (recombinant) (10µg)
SDP
Catalog No. 502013973
Encompass_Preferred
Alternative name: Interleukin-1α, Hematopoietin-1, Lymphocyte-activating factor , LAF, Endogenous pyrogen, Leukocyte endogenous mediator, LEM, Mononuclear cell factor, MCF, IL-1F1. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized from 10mM TRIS, pH 8.0, containing 50mM NaCl. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-3973 Supplier Enzo Life Sciences Supplier No. ENZPRT1860010
Alternative name: Interleukin-1α, Hematopoietin-1, Lymphocyte-activating factor , LAF, Endogenous pyrogen, Leukocyte endogenous mediator, LEM, Mononuclear cell factor, MCF, IL-1F1. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized from 10mM TRIS, pH 8.0, containing 50mM NaCl. Source: Produced in E. coli. Long Term Storage: -20°C.