Learn More
Enzo Life Sciences IL-1alpha (mouse), (recombinant) (10µg)
SDP
Catalog No. 502013974
Encompass_Preferred
Alternative name: Interleukin-1α, Hematopoietin-1, Lymphocyte-activating factor , LAF, Endogenous pyrogen, Leukocyte endogenous mediator, LEM, Mononuclear cell factor, MCF, IL-1F1. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized from 1mM TRIS, pH 8.0. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-3974 Supplier Enzo Life Sciences Supplier No. ENZPRT1870010
Alternative name: Interleukin-1α, Hematopoietin-1, Lymphocyte-activating factor , LAF, Endogenous pyrogen, Leukocyte endogenous mediator, LEM, Mononuclear cell factor, MCF, IL-1F1. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized from 1mM TRIS, pH 8.0. Source: Produced in E. coli. Long Term Storage: -20°C.