Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0311510
Encompass
Interleukin-1 alpha (IL-1Alpha) is produced in a variety of cells including monocytes tissue macrophages keratinocytes and other epithelial cells. Both IL-1 alpha and IL-1beta bind to the same receptor and have similar if not identical biological properties. These cytokines have a broad range of activities including stimulation of thymocyte proliferation via IL-2 release B-cell maturation and proliferation mitogenic FGF-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However whereas IL-1beta is a secreted cytokine IL-1 alpha is predominantly a cell-associated cytokine.
Catalog No. 50-252-9818