Learn More
CPC Scientific Ac-Tyr-Val-Lys-Asp-CHO 1MG
SDP
Catalog No. 502108948
Encompass
Affinity ligand for interleukin-1b converting enzyme. Slow binding caspase-1 (ICE) inhibitor with low selectivity.SEQUENCE: Ac-Tyr-Val-Lys-Asp-CHOONE-LETTER SEQUENCE: Ac-YVKD-CHOMOLECULAR FORMULA: C26H39N5O8MOLECULAR WEIGHT: 549.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [147821-01-1]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:T.L. Graybill et al., Int. J. Peptide Protein Res., 44, 173 (1994)N.A. Thornbery et al., Nature, 356, 768 (1992)SKU(s): CASP-015A
Catalog No. 50-210-8948 Supplier CPC Scientific Supplier No. CASP015A
Affinity ligand for interleukin-1b converting enzyme. Slow binding caspase-1 (ICE) inhibitor with low selectivity.SEQUENCE: Ac-Tyr-Val-Lys-Asp-CHOONE-LETTER SEQUENCE: Ac-YVKD-CHOMOLECULAR FORMULA: C26H39N5O8MOLECULAR WEIGHT: 549.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [147821-01-1]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:T.L. Graybill et al., Int. J. Peptide Protein Res., 44, 173 (1994)N.A. Thornbery et al., Nature, 356, 768 (1992)SKU(s): CASP-015A