Learn More
Enzo Life Sciences IL-1 beta (soluble) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences ALX5220563010
Encompass_Preferred
Alternative name: IL-1F2, Interleukin-1 beta. Concentration: 0.1mg/ml after reconstitution. Endotoxin content: E. coli. Human IL-1β (interleukin-1β) (aa 117-270) is fused to a linker peptide (10 aa) and an N-terminal FLAG-tag. Specificity: Binds to human IL-1R (interleukin-1 receptor). Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: P01584. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC1648239