Learn More
Medchemexpress LLC HY-P7073A 10ug Medchemexpress, IL-1beta (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934316
Encompass_Preferred
Medchemexpress, HY-P7073A 10ug IL-1beta (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-1 beta (CHO-expressed) is a CHO cell derived proinflammatory, cell-associated cytokine, produced by monocytes, tissue macrophages, keratinocytes and other epithelial cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4316 Supplier Medchemexpress LLC Supplier No. HYP7073A10UG
Medchemexpress, HY-P7073A 10ug IL-1beta (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-1 beta (CHO-expressed) is a CHO cell derived proinflammatory, cell-associated cytokine, produced by monocytes, tissue macrophages, keratinocytes and other epithelial cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.