Learn More
AdipoGen IL-1beta (human) (rec.) (untagged) (10 µg)
SDP
Supplier:  AdipoGen AG40B0023C010
Encompass_Preferred
Protein. Human IL-1beta (aa 117-269) is untagged. Source: E. coli. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human IL-1R. Purity: >95% (SDS-PAGE). IL-1beta is produced by activated macrophages. IL-1beta stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1beta belongs to the IL-1 family of proteins that are involved in the inflammatory response. IL-1beta in most situations requires the inflammasome complex to be cleaved and secreted.
Catalog No. 50-205-3094