Learn More
Enzo Life Sciences IL-1beta (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900130A
Encompass_Preferred
The IL-1β (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Quantify 1 pg/mL IL-1β with lot sample volume needed saving you precious sample. Alternative name: Interleukin-1beta. Assay Time: < 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-6850