Learn More
Medchemexpress LLC HY-P7073 10ug Medchemexpress, IL-1beta, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934315
Encompass_Preferred
Medchemexpress, HY-P7073 10ug IL-1beta, Mouse CAS: Recombinant Mouse Interleukin-1 beta is a proinflammatory, cell-associated cytokine, produced by monocytes, tissue macrophages, keratinocytes and other epithelial cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4315 Supplier Medchemexpress LLC Supplier No. HYP707310UG
Medchemexpress, HY-P7073 10ug IL-1beta, Mouse CAS: Recombinant Mouse Interleukin-1 beta is a proinflammatory, cell-associated cytokine, produced by monocytes, tissue macrophages, keratinocytes and other epithelial cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.