Learn More
Enzo Life Sciences IL-1beta (mouse), (recombinant) (10µg)
SDP
Catalog No. 502013928
Encompass_Preferred
Alternative name: Interleukin-1beta, Catabolin. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10 mM sodium citrate, pH 4.0. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3928 Supplier Enzo Life Sciences Supplier No. ENZPRT1360010
Alternative name: Interleukin-1beta, Catabolin. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 10 mM sodium citrate, pH 4.0. Source: Produced in E.coli. Long Term Storage: -20°C.