Learn More
ACROBiosystems ACROBiosystems Human IL-1 beta / IL-1F2 Protein, Tag Free, low endotoxin
Supplier:  ACROBiosystems ILBH411020UG
Encompass_Preferred
Recombinant Protein;20UG;Human IL-1 beta, Tag Free (ILB-H4110) is expressed from E.coli cells. It contains AA Ala 117 - Ser 269 (Accession # NP_000567).;This protein carries no "tag". The protein has a calculated MW of 17.5 kDa. The protein migrates as 18 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IL-1 beta, Tag Free (Cat. No. ILB-H4110) at 10 μg/mL (100 μL/well) can bind Human IL-1 RII, Fc Tag (Cat. No. IL2-H4256) with a linear range of 4-125 ng/mL (QC tested).
Catalog No. 50-195-6530