Learn More
Medchemexpress LLC IL-1RA/IL-1F3 Porci 500ug | 500ug
SDP
Catalog No. 5000195262
Encompass_Preferred
IL-1RA/IL-1F3 proteins are powerful anti-inflammatory antagonists in the interleukin 1 family specifically targeting the pro-inflammatory cytokines IL1B and IL1A This protein counteracts the inflammatory effects of IL1 playing a critical role in preventing immune dysregulation and preventing uncontrolled systemic inflammation triggered by a variety of innate irritants including pathogens IL-1RA/IL-1F3 Protein Porcine is the recombinant Porcine-derived IL-1RA/IL-1F3 protein expressed by E coli with tag free
Catalog No. 50-001-95262 Supplier Medchemexpress LLC Supplier No. HYP79293500UG
IL-1RA/IL-1F3 proteins are powerful anti-inflammatory antagonists in the interleukin 1 family specifically targeting the pro-inflammatory cytokines IL1B and IL1A This protein counteracts the inflammatory effects of IL1 playing a critical role in preventing immune dysregulation and preventing uncontrolled systemic inflammation triggered by a variety of innate irritants including pathogens IL-1RA/IL-1F3 Protein Porcine is the recombinant Porcine-derived IL-1RA/IL-1F3 protein expressed by E coli with tag free