Learn More
Medchemexpress LLC IL-1RAcP/IL-1 R3 Mo 100ug | 100ug
SDP
Catalog No. 5000186772
Encompass_Preferred
IL-1RAcP is a co-receptor for IL1RL2 IL1R1 and IL1RL1 in the IL-36 IL-1 and IL-33 signaling system respectively IL-1RAcP is an essential component of the IL-1 receptor and plays an essential role in the inflammatory process[1][2] IL-1RAcP/IL-1 R3 Protein Mouse (HEK293 Fc) is a recombinant mouse IL-1RAcP protein with an hFc tag at the C-terminus and is expressed in HEK293 cells
Catalog No. 50-001-86772 Supplier Medchemexpress LLC Supplier No. HYP77417100UG
IL-1RAcP is a co-receptor for IL1RL2 IL1R1 and IL1RL1 in the IL-36 IL-1 and IL-33 signaling system respectively IL-1RAcP is an essential component of the IL-1 receptor and plays an essential role in the inflammatory process[1][2] IL-1RAcP/IL-1 R3 Protein Mouse (HEK293 Fc) is a recombinant mouse IL-1RAcP protein with an hFc tag at the C-terminus and is expressed in HEK293 cells