Learn More
ACROBiosystems ACROBiosystems Human IL-1RL1 / ST2 Protein, Fc Tag
Catalog No. 501957165
Encompass_Preferred
Recombinant Protein;1MG;Human IL-1RL1, Fc Tag (IL1-H5250) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Pro 323 (Accession # NP_003847).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 61.0 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) at 5 μg/mL (100 μL/well) can bind Human IL-33, His Tag (Cat. No. IL3-H52H7) with a linear range of 5-40 ng/mL (QC tested).
Catalog No. 50-195-7165 Supplier ACROBiosystems Supplier No. IL1H52501MG
Recombinant Protein;1MG;Human IL-1RL1, Fc Tag (IL1-H5250) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Pro 323 (Accession # NP_003847).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 61.0 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) at 5 μg/mL (100 μL/well) can bind Human IL-33, His Tag (Cat. No. IL3-H52H7) with a linear range of 5-40 ng/mL (QC tested).