Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-1RN Horse His 100ug
SDP

Catalog No. 5000196120
Encompass_Preferred

Recombinant equine interleukin-1 receptor antagonist (IL-1RN/IL-1ra) supplied as a lyophilized, N-6xHis-tagged protein for research use. The product is provided as 100 μg of protein with confirmed purity and biological activity, formulated for stability and long-term storage.

  • Recombinant equine interleukin-1 receptor antagonist with N-6xHis tag.
  • Expressed in E. coli and lyophilized for stability.
  • Supplied as 100 μg of protein, approximately 18 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Demonstrated biological activity: ED50 3.666 μg/mL (specific activity 272.78 units/mg).
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4 formulation.
  • Store at -20°C; aliquot to avoid freeze-thaw for long-term storage.

Catalog No. 50-001-96120 Supplier Medchemexpress LLC Supplier No. HYP71876A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant equine interleukin-1 receptor antagonist (IL-1RN/IL-1ra) supplied as a lyophilized, N-6xHis-tagged protein for research use. The product is provided as 100 μg of protein with confirmed purity and biological activity, formulated for stability and long-term storage.

  • Recombinant equine interleukin-1 receptor antagonist with N-6xHis tag.
  • Expressed in E. coli and lyophilized for stability.
  • Supplied as 100 μg of protein, approximately 18 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Demonstrated biological activity: ED50 3.666 μg/mL (specific activity 272.78 units/mg).
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4 formulation.
  • Store at -20°C; aliquot to avoid freeze-thaw for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.