Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-1RN Horse His 10ug
SDP

Catalog No. 5000196122
Encompass_Preferred

Recombinant IL-1RN protein from horse (equine), expressed in Escherichia coli and supplied lyophilized with an N-terminal 6xHis tag. Intended for research on IL-1 signaling and inflammation, the product has a predicted molecular mass of ~17.2 kDa and a purity greater than 95% by reducing SDS-PAGE. Reconstitution guidance advises concentrations ≥100 μg/mL; store lyophilized at -20°C and freeze aliquots at -20°C or -80°C for extended storage.

  • Recombinant equine interleukin-1 receptor antagonist with N-terminal 6xHis tag.
  • Expressed in Escherichia coli.
  • Predicted molecular mass ~17.2 kDa (≈18 kDa by SDS-PAGE).
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Reconstitution recommended at concentrations ≥100 μg/mL.
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C for long term.

Catalog No. 50-001-96122 Supplier Medchemexpress LLC Supplier No. HYP71876A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant IL-1RN protein from horse (equine), expressed in Escherichia coli and supplied lyophilized with an N-terminal 6xHis tag. Intended for research on IL-1 signaling and inflammation, the product has a predicted molecular mass of ~17.2 kDa and a purity greater than 95% by reducing SDS-PAGE. Reconstitution guidance advises concentrations ≥100 μg/mL; store lyophilized at -20°C and freeze aliquots at -20°C or -80°C for extended storage.

  • Recombinant equine interleukin-1 receptor antagonist with N-terminal 6xHis tag.
  • Expressed in Escherichia coli.
  • Predicted molecular mass ~17.2 kDa (≈18 kDa by SDS-PAGE).
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Reconstitution recommended at concentrations ≥100 μg/mL.
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.