Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-1RN Horse His 2ug
SDP

Catalog No. 5000196215
Encompass_Preferred

IL-1RN Protein, Horse (His) is a recombinant equine interleukin-1 receptor antagonist produced in Escherichia coli and supplied as a lyophilized, His-tagged protein for in vitro biochemical and binding studies. The preparation is highly pure and formulated for convenient storage and reconstitution to support downstream assays.

  • Recombinant equine IL-1RN with N-terminal His tag.
  • Expressed in Escherichia coli for efficient production.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass ~17.2 kDa (~18 kDa on SDS-PAGE).
  • Lyophilized formulation for convenient storage and transport.
  • Buffered formulation; buffer exchange recommended for sensitive assays such as SPR.
  • Stable under recommended storage: -20°C for long term; reconstituted stable at 4°C for 1 week.
  • Suitable for biochemical, binding, and in vitro functional assays.
  • Sequence corresponds to residues H26-Q177 of equine IL-1RN.

Catalog No. 50-001-96215 Supplier Medchemexpress LLC Supplier No. HYP71876A2UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IL-1RN Protein, Horse (His) is a recombinant equine interleukin-1 receptor antagonist produced in Escherichia coli and supplied as a lyophilized, His-tagged protein for in vitro biochemical and binding studies. The preparation is highly pure and formulated for convenient storage and reconstitution to support downstream assays.

  • Recombinant equine IL-1RN with N-terminal His tag.
  • Expressed in Escherichia coli for efficient production.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass ~17.2 kDa (~18 kDa on SDS-PAGE).
  • Lyophilized formulation for convenient storage and transport.
  • Buffered formulation; buffer exchange recommended for sensitive assays such as SPR.
  • Stable under recommended storage: -20°C for long term; reconstituted stable at 4°C for 1 week.
  • Suitable for biochemical, binding, and in vitro functional assays.
  • Sequence corresponds to residues H26-Q177 of equine IL-1RN.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.