Learn More
Medchemexpress LLC IL-1RN Horse His 50ug
SDP
Catalog No. 5000195743
Encompass_Preferred
Recombinant equine interleukin-1 receptor antagonist (IL-1RN/IL-1RA) expressed in E. coli and supplied as a lyophilized, N-terminal 6xHis-tagged protein. It is intended for research into IL-1 signaling and inflammatory processes, and is provided with formulation, purity, and storage information to support experimental use.
  • Expressed in E. coli with an N-terminal 6xHis tag.
  • Derived from equine (horse) sequence (UniProt O18999, residues H26-Q177).
  • Predicted mass 17.2 kDa; observed ~18 kDa by SDS-PAGE.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Store lyophilized at -20°C; after reconstitution, aliquot and store at -20°C or -80°C for long term.
  • Available in multiple sizes including 50 μg and 100 μg.
Catalog No. 50-001-95743 Supplier Medchemexpress LLC Supplier No. HYP71876A50UG
Recombinant equine interleukin-1 receptor antagonist (IL-1RN/IL-1RA) expressed in E. coli and supplied as a lyophilized, N-terminal 6xHis-tagged protein. It is intended for research into IL-1 signaling and inflammatory processes, and is provided with formulation, purity, and storage information to support experimental use.
  • Expressed in E. coli with an N-terminal 6xHis tag.
  • Derived from equine (horse) sequence (UniProt O18999, residues H26-Q177).
  • Predicted mass 17.2 kDa; observed ~18 kDa by SDS-PAGE.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Store lyophilized at -20°C; after reconstitution, aliquot and store at -20°C or -80°C for long term.
  • Available in multiple sizes including 50 μg and 100 μg.