Learn More
ACROBiosystems ACROBiosystems Human IL-2 Protein, His Tag
Catalog No. 501956463
Encompass_Preferred
Recombinant Protein;1MG;Human IL-2, His Tag (IL2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession # P60568-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.3 kDa. The protein migrates as 18 kDa and 19 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2 R alpha, Fc Tag (Cat. No. ILA-H5251) at 5 μg/mL (100 μL/well) can bind Human IL-2, His Tag (Cat. No. IL2-H52H8) with a linear range of 0.3-2.5 ng/mL (QC tested).
Catalog No. 50-195-6463 Supplier ACROBiosystems Supplier No. IL2H52H81MG
Recombinant Protein;1MG;Human IL-2, His Tag (IL2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession # P60568-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.3 kDa. The protein migrates as 18 kDa and 19 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2 R alpha, Fc Tag (Cat. No. ILA-H5251) at 5 μg/mL (100 μL/well) can bind Human IL-2, His Tag (Cat. No. IL2-H52H8) with a linear range of 0.3-2.5 ng/mL (QC tested).