Learn More
Enzo Life Sciences IL-2 (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2540100
Encompass_Preferred
Alternative name: Interleukin-2, T cell growth factor, TCGF, Aldesleukin. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 134 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4070