Learn More
Sigma Aldrich Fine Chemicals Biosciences IL 2 human Animal componen
Catalog No. 502905028
Encompass
IL-2 (interleukin 2) is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. It is a type I cytokine. The gene is mapped to human chromosome 4q27. Recombinant human IL-2 is a 15.5kDa protein containing 134 amino acid residues including one intrachain disulfide bond.
Catalog No. 50-290-5028 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP308550UG
IL-2 (interleukin 2) is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. It is a type I cytokine. The gene is mapped to human chromosome 4q27. Recombinant human IL-2 is a 15.5kDa protein containing 134 amino acid residues including one intrachain disulfide bond.