Learn More
BPS Bioscience Inc IL-2 Luciferase Reporter Jurkat Cell Line, Size: 2 vials
SDP
Catalog No. NC3920802
Encompass
This recombinant Jurkat cell line was engineered for conditional expression of firefly luciferase. It contains a firefly luciferase gene under the control of a human Interleukin-2 (IL-2) promoter stably integrated into the genome.The IL-2 Luciferase Reporter Jurkat cell line is suitable to monitor the activation of the IL-2 promoter. This reporter cell line has been functionally validated using various agonists such as phorbol 12-myristate 13-acetate (PMA) in combination with ionomycin, an anti-human CD3 antibody, anti-human CD3/CD28 Dynabeads, and an anti-CD19/anti-CD3 bispecific antibody. It has also been validated in a co-culture assay with our TCR activator CHO cell line.
Catalog No. NC3920802 Supplier BPS Bioscience Inc Supplier No. 60481
This recombinant Jurkat cell line was engineered for conditional expression of firefly luciferase. It contains a firefly luciferase gene under the control of a human Interleukin-2 (IL-2) promoter stably integrated into the genome.The IL-2 Luciferase Reporter Jurkat cell line is suitable to monitor the activation of the IL-2 promoter. This reporter cell line has been functionally validated using various agonists such as phorbol 12-myristate 13-acetate (PMA) in combination with ionomycin, an anti-human CD3 antibody, anti-human CD3/CD28 Dynabeads, and an anti-CD19/anti-CD3 bispecific antibody. It has also been validated in a co-culture assay with our TCR activator CHO cell line.