Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z032971
Encompass
Interleukin-2 receptor (IL-2R) is a heterotrimeric protein expressed on the surface of certain immune cells such as lymphocytes that binds and responds to the cytokine IL-2. The IL-2R is made up of 3 subunits - alpha (Alpha) beta (beta ) and gamma (gamma). The Alpha andBeta chains are involved in binding IL-2 while signal transduction following cytokine interaction is carried out by the gamma-chain along with theBeta subunit. TheBeta and gamma chains of the IL-2R are members of the type I cytokine receptor family. IL-2R has a high binding affinity to IL-2 and is expressed by antigen-activated T lymphocytes (T cells). IL-2 RAlpha is also known as CD25 p55 and Tac (activated T cell) antigen.
Catalog No. 50-252-9687