Learn More
ACROBiosystems Mouse IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag (ILG-M5254) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 240 (IL-2RB) & Trp 23 - Ala 263 (IL-2RG) (Accession # P16297-1 (IL-2RB) & P34902-1 (IL-2RG)).
Catalog No. 502230765
Encompass_Preferred
Both Interleukin-2 receptor subunit beta and Interleukin-2 receptor subunit gamma are receptor for interleukin-2. Common subunit for the receptors for a variety of interleukins. Interacts with SHB upon interleukin stimulation. Probably in association with IL15RA, involved in the stimulation of neutrophil phagocytosis by IL15. This beta subunit is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2. IL2R exists in 3 different forms: a high affinity dimer, an intermediate affinity monomer (beta subunit), and a low affinity monomer (alpha subunit). The high and intermediate affinity forms also associate with a gamma subunit.
This item is not returnable. View return policy
Catalog No. 50-223-0765 Supplier ACROBiosystems Supplier No. ILGM5254100UG
Both Interleukin-2 receptor subunit beta and Interleukin-2 receptor subunit gamma are receptor for interleukin-2. Common subunit for the receptors for a variety of interleukins. Interacts with SHB upon interleukin stimulation. Probably in association with IL15RA, involved in the stimulation of neutrophil phagocytosis by IL15. This beta subunit is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2. IL2R exists in 3 different forms: a high affinity dimer, an intermediate affinity monomer (beta subunit), and a low affinity monomer (alpha subunit). The high and intermediate affinity forms also associate with a gamma subunit.