Learn More
Medchemexpress LLC IL-2 Rat HEK293 H 10ug
SDP
Catalog No. 5000185449
Encompass_Preferred
Recombinant rat interleukin-2 produced in HEK293 cells and supplied as a C-terminal 6xHis-tagged protein for in vitro immunology and cell-based assays. The product is provided as a 0.22 μm filtered solution with low endotoxin and high purity, enabling reliable activity measurements and assay development in research settings.
  • Rat species recombinant interleukin-2 for species-relevant assays.
  • Expressed in HEK293 cells to support mammalian post-translational modifications.
  • C-terminal 6xHis tag enables straightforward detection and purification.
  • High purity (>95%) by reducing SDS-PAGE for consistent performance.
  • Low endotoxin (<1 EU/μg) suitable for sensitive cell-based assays.
  • Supplied as a 0.22 μm filtered solution formulated at pH 4.0.
  • Validated bioactivity with ED50 approximately 12.7-16.3 ng/mL in CTLL-2 proliferation assay.
  • Available in small research sizes for assay development and validation.
Catalog No. 50-001-85449 Supplier Medchemexpress LLC Supplier No. HYP7071810UG
Recombinant rat interleukin-2 produced in HEK293 cells and supplied as a C-terminal 6xHis-tagged protein for in vitro immunology and cell-based assays. The product is provided as a 0.22 μm filtered solution with low endotoxin and high purity, enabling reliable activity measurements and assay development in research settings.
  • Rat species recombinant interleukin-2 for species-relevant assays.
  • Expressed in HEK293 cells to support mammalian post-translational modifications.
  • C-terminal 6xHis tag enables straightforward detection and purification.
  • High purity (>95%) by reducing SDS-PAGE for consistent performance.
  • Low endotoxin (<1 EU/μg) suitable for sensitive cell-based assays.
  • Supplied as a 0.22 μm filtered solution formulated at pH 4.0.
  • Validated bioactivity with ED50 approximately 12.7-16.3 ng/mL in CTLL-2 proliferation assay.
  • Available in small research sizes for assay development and validation.