Learn More
AdipoGen IL-2 SuperkH9T MonoFc-KIHh
SDP
Catalog No. 502562846
Encompass_Preferred
Recombinant Protein. The extracellular domain of human IL-2 aa 21-153 mutant H9T containing the mutations of H9 L80F / R81D / L85V / I 86V / I92F and the new mutation Q126T is fused at the C-terminus to the Fc portion of human IgG1 Knobs-into-Holes technology see reference J.B. Ridgway, et al. Protein Eng. 9, 617 1996. Source HEK 293 cells. Lyophilized. Contains PBS. Interleukin-2 IL-2 is a 133 amino acid glycoprotein with one intramolecular disulfide bond and variable glycosylation. It is secreted by activated T cells and induces proliferation and maturation of activated T cells, natural killer cells and lymphokine activated killer cells. IL-2 also stimulates proliferation of antibody-producing B cells, activates neutrophils and induces mononuclear cells to secrete IFN-gamma and TNF-alpha and -beta.
Catalog No. 50-256-2846 Supplier AdipoGen Supplier No. AG40B0223C100
Recombinant Protein. The extracellular domain of human IL-2 aa 21-153 mutant H9T containing the mutations of H9 L80F / R81D / L85V / I 86V / I92F and the new mutation Q126T is fused at the C-terminus to the Fc portion of human IgG1 Knobs-into-Holes technology see reference J.B. Ridgway, et al. Protein Eng. 9, 617 1996. Source HEK 293 cells. Lyophilized. Contains PBS. Interleukin-2 IL-2 is a 133 amino acid glycoprotein with one intramolecular disulfide bond and variable glycosylation. It is secreted by activated T cells and induces proliferation and maturation of activated T cells, natural killer cells and lymphokine activated killer cells. IL-2 also stimulates proliferation of antibody-producing B cells, activates neutrophils and induces mononuclear cells to secrete IFN-gamma and TNF-alpha and -beta.