Learn More
Enzo Life Sciences IL-20 (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2470010
Encompass_Preferred
Alternative name: Interleukin-20, Cytokine Zcyto10. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium phosphate, pH 7.5. Source: Produced in E. coli. Contains 153/306 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4058